Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF384 SIRNA Assays ›
Gene Symbol:
ZNF384
Gene Name:
zinc finger protein 384
Gene Aliases:
CAGH1, CAGH1A, CIZ, ERDA2, NMP4, NP, TNRC1
Chromosome Location:
Chr.12: 6666477 - 6689572 on Build GRCh38
UniGene ID:
Hs.103315
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS176160
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF384
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039920.2
NP_001035009.1
8, 9
1253
NM_001135734.2
NP_001129206.1
10, 11
1601
NM_133476.4
NP_597733.2
9, 10
1491
XM_006718969.3
XP_006719032.1
6, 7
790
XM_011520917.1
XP_011519219.1
11, 12
1985
XM_011520918.1
XP_011519220.1
11, 12
1694
XM_011520919.1
XP_011519221.1
11, 12
1735
XM_011520920.1
XP_011519222.1
11, 12
1719
XM_011520921.2
XP_011519223.1
11, 12
1544
XM_011520922.1
XP_011519224.1
10, 11
1646
XM_011520923.1
XP_011519225.1
10, 11
1671
XM_017018941.1
XP_016874430.1
10, 11
1626
XM_017018942.1
XP_016874431.1
10, 11
1492
XM_017018943.1
XP_016874432.1
10, 11
1486
XM_017018944.1
XP_016874433.1
9, 10
1553
XM_017018945.1
XP_016874434.1
9, 10
1578
XM_017018946.1
XP_016874435.1
10, 11
1536
XM_017018947.1
XP_016874436.1
10, 11
1552
XM_017018948.1
XP_016874437.1
8, 9
1980
XM_017018949.1
XP_016874438.1
9, 10
1418
XM_017018950.1
XP_016874439.1
9, 10
1459
XM_017018951.1
XP_016874440.1
8, 9
1370
XM_017018952.1
XP_016874441.1
8, 9
1395
XM_017018953.1
XP_016874442.1
8, 9
1411
XM_017018954.1
XP_016874443.1
9, 10
2209
GenBank mRNA
AK095734.1
-
9, 10
1440
AK298962.1
-
10, 11
1539
BC053361.1
-
8, 9
1135
6012540
CR987433.1
-
4, 5
641
U80738.1
-
1, 2
151
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
nucleocytoplasmic transport
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein secretion
SH3 domain binding
sequence-specific DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline