Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCNY SIRNA Assays ›
          Gene Symbol:
          CCNY
          Gene Name:
          cyclin Y
          Gene Aliases:
          C10orf9, CBCP1, CCNX, CFP1
          Chromosome Location:
          Chr.10: 35246981 - 35572670 on Build GRCh38
          UniGene ID:
          Hs.14745
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS176729
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCNY
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282852.1 NP_001269781.1 6, 7 688
          NM_001282853.1 NP_001269782.1 8, 9 826
          NM_001282854.1 NP_001269783.1 5, 6 626
          NM_145012.5 NP_659449.3 7, 8 763
          NM_181698.3 NP_859049.2 9, 10 787
          XM_006717392.3 XP_006717455.1 7, 8 532
          XM_011519357.2 XP_011517659.1 6, 7 722
          XM_011519358.1 XP_011517660.1 10, 11 854
          XM_011519360.2 XP_011517662.1 11, 12 956
          XM_011519361.1 XP_011517663.1 11, 12 838
          XM_011519362.1 XP_011517664.1 10, 11 818
          XM_011519363.1 XP_011517665.1 10, 11 831
          XM_011519364.1 XP_011517666.1 12, 13 952
          XM_011519365.2 XP_011517667.1 11, 12 871
          XM_011519366.1 XP_011517668.1 10, 11 801
          XM_011519367.1 XP_011517669.1 8, 9 631
          XM_011519368.2 XP_011517670.1 7, 8 526
          XM_011519369.2 XP_011517671.1 6, 7 1016
          XM_017015839.1 XP_016871328.1 7, 8 449
          XM_017015840.1 XP_016871329.1 7, 8 451
          XM_017015841.1 XP_016871330.1 7, 8 449
          GenBank mRNA AF413522.1 - 7, 8 763
          AF429969.1 - 9, 10 743
          AF465728.1 - 7, 8 509
          AF465729.1 - 7, 8 584
          AK057280.1 - 9, 10 743
          AK303283.1 - 5, 6 480
          AK314666.1 - 8, 9 802
          AL834355.1 - 7, 8 621
          AY504868.1 - 7, 8 763
          BC069224.1 - 7, 8 589
          BC094815.1 - 7, 8 615 6340750
          BC104773.1 - 8, 9 572 8143776
          BC104801.1 - 8, 9 572 8143804
          BC143450.1 - 7, 8 465 9051958
          BC143455.1 - 8, 9 572 9051963

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          Wnt signaling pathway
          positive regulation of cyclin-dependent protein serine/threonine kinase activity
          cell division
          regulation of canonical Wnt signaling pathway
          protein binding
          cyclin-dependent protein serine/threonine kinase regulator activity
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix