Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FGFR1 SIRNA Assays ›
          Gene Symbol:
          FGFR1
          Gene Name:
          fibroblast growth factor receptor 1
          Gene Aliases:
          BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
          Chromosome Location:
          Chr.8: 38411138 - 38468834 on Build GRCh38
          UniGene ID:
          Hs.264887
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS176921
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FGFR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001174063.1 NP_001167534.1 7 1723
          NM_001174064.1 NP_001167535.1 8 1791
          NM_001174065.1 NP_001167536.1 7 1037
          NM_001174066.1 NP_001167537.1 6 776
          NM_001174067.1 NP_001167538.1 8 1197
          NM_015850.3 NP_056934.2 7 1717
          NM_023105.2 NP_075593.1 6 1456
          NM_023106.2 NP_075594.1 6 1450
          NM_023110.2 NP_075598.2 7 1723
          XM_006716303.2 XP_006716366.1 7 1524
          XM_006716304.1 XP_006716367.1 7 1040
          XM_006716306.2 XP_006716369.1 7 1518
          XM_006716307.1 XP_006716370.1 7 1034
          XM_006716309.3 XP_006716372.1 7 1494
          XM_006716310.2 XP_006716373.1 6 1257
          XM_006716311.1 XP_006716374.1 6 655
          XM_006716312.1 XP_006716375.1 6 725
          XM_006716313.2 XP_006716376.1 6 1251
          XM_006716314.1 XP_006716377.1 6 729
          XM_011544443.2 XP_011542745.1 7 1029
          XM_011544444.1 XP_011542746.1 7 1018
          XM_011544445.2 XP_011542747.1 7 1037
          XM_011544446.2 XP_011542748.1 7 1024
          XM_011544447.2 XP_011542749.1 7 1024
          XM_011544448.1 XP_011542750.1 6 628
          XM_011544449.1 XP_011542751.1 6 622
          XM_011544450.2 XP_011542752.1 6 622
          XM_011544451.1 XP_011542753.1 5 491
          XM_011544452.2 XP_011542754.1 7 1112
          XM_017013219.1 XP_016868708.1 7 1025
          XM_017013220.1 XP_016868709.1 7 1018
          XM_017013221.1 XP_016868710.1 8 1233
          XM_017013222.1 XP_016868711.1 7 1524
          XM_017013223.1 XP_016868712.1 7 1034
          XM_017013224.1 XP_016868713.1 7 1518
          XM_017013225.1 XP_016868714.1 7 1001
          XM_017013226.1 XP_016868715.1 6 628
          XM_017013227.1 XP_016868716.1 6 622
          XM_017013228.1 XP_016868717.1 6 1251
          XM_017013230.1 XP_016868719.1 5 582
          XM_017013231.1 XP_016868720.1 7 1122
          GenBank mRNA AB208919.1 - 8 1578
          AK222718.1 - 7 1516
          AK223126.1 - 6 1251
          AK291754.1 - 7 916
          AK292470.1 - 6 776
          AK309947.1 - 8 1076
          BC015035.1 - 8 1451 3911101
          BC018128.1 - 7 1481 3896359
          BC091494.1 - 5 587 30387634
          BX649167.1 - 3 3065
          EU826596.1 - 5 508
          EU826597.1 - 5 514
          FJ809916.1 - 7 1726
          FJ809917.1 - 7 1726
          KU178066.1 - 6 781
          KU178067.1 - 5 514
          KU178068.1 - 5 508
          KU178069.1 - 5 514
          KU178070.1 - 5 508
          M34185.1 - 6 769
          M34186.1 - 6 1034
          M34187.1 - 6 930
          M34188.1 - 6 1030
          M34641.1 - 6 784
          M37722.1 - 6 736
          M60485.1 - 8 1446
          M63887.1 - 6 781
          M63888.1 - 6 781
          M63889.1 - 6 658
          X51803.1 - 7 898
          X52833.1 - 6 868
          X57119.1 - 6 676
          X57120.1 - 7 937
          X57121.1 - 7 943
          X57122.1 - 6 670
          X66945.1 - 7 1507
          Y00665.1 - 3 181

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          skeletal system development
          angiogenesis
          ureteric bud development
          in utero embryonic development
          organ induction
          neuron migration
          positive regulation of mesenchymal cell proliferation
          chondrocyte differentiation
          transcription, DNA-templated
          protein phosphorylation
          sensory perception of sound
          positive regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          positive regulation of phospholipase activity
          positive regulation of phospholipase C activity
          positive regulation of neuron projection development
          regulation of phosphatidylinositol 3-kinase signaling
          positive regulation of phosphatidylinositol 3-kinase signaling
          cell migration
          peptidyl-tyrosine phosphorylation
          stem cell population maintenance
          motogenic signaling involved in postnatal olfactory bulb interneuron migration
          ventricular zone neuroblast division
          embryonic limb morphogenesis
          midbrain development
          neuron projection development
          fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
          phosphatidylinositol-3-phosphate biosynthetic process
          inner ear morphogenesis
          outer ear morphogenesis
          middle ear morphogenesis
          chordate embryonic development
          positive regulation of MAP kinase activity
          positive regulation of MAPK cascade
          positive regulation of GTPase activity
          regulation of cell differentiation
          positive regulation of neuron differentiation
          negative regulation of osteoblast differentiation
          positive regulation of cell cycle
          protein autophosphorylation
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          paraxial mesoderm development
          regulation of lateral mesodermal cell fate specification
          cell maturation
          skeletal system morphogenesis
          mesenchymal cell differentiation
          regulation of sensory perception of pain
          positive regulation of cardiac muscle cell proliferation
          auditory receptor cell development
          branching involved in salivary gland morphogenesis
          lung-associated mesenchyme development
          regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
          regulation of stem cell proliferation
          positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
          positive regulation of endothelial cell chemotaxis to fibroblast growth factor
          regulation of extrinsic apoptotic signaling pathway in absence of ligand
          glycoprotein binding
          protein tyrosine kinase activity
          fibroblast growth factor-activated receptor activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          ATP binding
          heparin binding
          1-phosphatidylinositol-3-kinase activity
          fibroblast growth factor binding
          protein complex binding
          identical protein binding
          protein homodimerization activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity
          cell adhesion molecule binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix