Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CARD8 SIRNA Assays ›
Gene Symbol:
CARD8
Gene Name:
caspase recruitment domain family member 8
Gene Aliases:
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN
Chromosome Location:
Chr.19: 48203148 - 48256269 on Build GRCh38
UniGene ID:
Hs.446146
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS177013
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CARD8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184900.1
NP_001171829.1
7
906
NM_001184901.1
NP_001171830.1
7
853
NM_001184902.1
NP_001171831.1
10
1269
NM_001184903.1
NP_001171832.1
8
1003
NM_014959.3
NP_055774.2
8
964
NR_033678.1
6
823
NR_033679.1
6
865
NR_033680.1
6
823
XM_006723091.3
XP_006723154.1
9
1117
XM_006723092.3
XP_006723155.1
11
1655
XM_006723093.3
XP_006723156.1
9
1438
XM_006723094.3
XP_006723157.1
9
1097
XM_006723095.3
XP_006723158.1
9
1084
XM_006723096.3
XP_006723159.1
10
1506
XM_006723097.3
XP_006723160.1
8
1289
XM_006723098.3
XP_006723161.1
8
1076
XM_006723099.3
XP_006723162.1
7
962
XM_006723101.2
XP_006723164.1
8
979
XM_006723102.3
XP_006723165.1
7
919
XM_006723104.3
XP_006723167.1
8
976
XM_006723106.3
XP_006723169.1
9
1449
XM_006723107.3
XP_006723170.1
9
1100
XM_006723109.3
XP_006723172.1
8
976
XM_011526641.2
XP_011524943.1
12
1962
XM_011526642.2
XP_011524944.1
11
1857
XM_011526643.2
XP_011524945.1
10
1544
XM_011526644.2
XP_011524946.1
8
1129
XM_011526646.2
XP_011524948.1
9
1603
XM_011526647.2
XP_011524949.1
8
1492
XM_011526648.2
XP_011524950.1
9
1609
XM_011526650.2
XP_011524952.1
9
1100
XM_011526653.2
XP_011524955.1
9
1639
XM_017026482.1
XP_016881971.1
9
1442
XM_017026483.1
XP_016881972.1
9
1395
XM_017026484.1
XP_016881973.1
9
1403
XM_017026485.1
XP_016881974.1
9
1100
XM_017026486.1
XP_016881975.1
7
959
XM_017026487.1
XP_016881976.1
6
1328
XM_017026488.1
XP_016881977.1
10
1544
XM_017026489.1
XP_016881978.1
9
1129
XM_017026490.1
XP_016881979.1
9
1100
XM_017026491.1
XP_016881980.1
9
1100
XM_017026492.1
XP_016881981.1
9
1411
XM_017026493.1
XP_016881982.1
8
1303
XM_017026494.1
XP_016881983.1
9
1917
XM_017026495.1
XP_016881984.1
8
1059
XM_017026496.1
XP_016881985.1
8
979
XM_017026497.1
XP_016881986.1
7
919
XM_017026498.1
XP_016881987.1
7
916
XM_017026499.1
XP_016881988.1
6
2689
GenBank mRNA
AB023172.1
-
6
858
AF322184.1
-
6
858
AF331519.1
-
4
545
AF405558.1
-
4
545
AF511652.1
-
6
858
AK297045.1
-
8
989
AK302327.1
-
6
793
AY026322.1
-
4
545
BX641091.1
-
3
776
BX648568.1
-
8
3010
EU118120.1
-
7
906
EU118121.1
-
6
865
EU118122.1
-
4
706
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of interleukin-1 beta secretion
protein binding
cysteine-type endopeptidase activator activity involved in apoptotic process
NACHT domain binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0