Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSWIM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242487.1 | NP_001229416.1 | 16 | 3329 | ||
NM_001242488.1 | NP_001229417.1 | 16 | 3329 | |||
NM_015037.3 | NP_055852.2 | 16 | 3344 | |||
XM_005269648.2 | XP_005269705.1 | 16 | 3391 | |||
XM_005269653.3 | XP_005269710.1 | 16 | 3089 | |||
XM_005269655.3 | XP_005269712.1 | 17 | 2987 | |||
XM_006717725.3 | XP_006717788.1 | 16 | 3376 | |||
XM_011539544.1 | XP_011537846.1 | 16 | 3391 | |||
XM_017015978.1 | XP_016871467.1 | 16 | 3376 | |||
GenBank mRNA | AB020720.2 | - | 17 | 3230 | ||
AK023742.1 | - | 7 | 898 | |||
AL133662.1 | - | 8 | 1926 | |||
BC111006.1 | - | 7 | 1092 | |||
BC117205.1 | - | 7 | 891 | |||
BC151206.1 | - | 16 | 3291 | 40147119 |
Gene Symbol: ZSWIM8-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038357.1 | 4 | 1141 | |||
GenBank mRNA | AL080182.1 | - | 4 | 1141 | ||
BQ003405.1 | - | 1 | 332 |