Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015150.1 | NP_055965.1 | 6, 7 | 1306 | ||
XM_005264985.1 | XP_005265042.1 | 6, 7 | 1263 | |||
XM_005264986.2 | XP_005265043.1 | 6, 7 | 1160 | |||
XM_006713069.1 | XP_006713132.1 | 6, 7 | 1169 | |||
XM_011533529.1 | XP_011531831.1 | 8, 9 | 1703 | |||
XM_011533530.1 | XP_011531832.1 | 6, 7 | 1331 | |||
XM_017006005.1 | XP_016861494.1 | 8, 9 | 1703 | |||
GenBank mRNA | AK074446.1 | - | 6, 7 | 1112 | ||
AK301024.1 | - | 5, 6 | 999 | |||
AK303846.1 | - | 2, 3 | 303 | |||
AY237654.1 | - | 6, 7 | 1297 | |||
BC006400.2 | - | 2, 3 | 234 | |||
BC014032.2 | - | 2, 3 | 234 | |||
BC032349.1 | - | 5, 6 | 979 | |||
BC051336.1 | - | 6, 7 | 1083 | |||
BC069209.1 | - | 6, 7 | 1083 | |||
BC098270.1 | - | 6, 7 | 1130 | 40001191 | ||
BC100667.1 | - | 6, 7 | 1130 | 40001188 | ||
BC100669.1 | - | 6, 7 | 1130 | 40001190 | ||
BC121795.1 | - | 6, 7 | 1130 | 40011545 | ||
D42043.1 | - | 6, 7 | 1234 |