Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FLT3LG SIRNA Assays ›
Gene Symbol:
FLT3LG
Gene Name:
fms related tyrosine kinase 3 ligand
Gene Aliases:
FL, FLT3L
Chromosome Location:
Chr.19: 49474172 - 49487038 on Build GRCh38
UniGene ID:
Hs.428
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS177277
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FLT3LG
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204502.1
NP_001191431.1
5
403
NM_001204503.1
NP_001191432.1
4
372
NM_001278637.1
NP_001265566.1
5
431
NM_001278638.1
NP_001265567.1
5
420
NM_001459.3
NP_001450.2
5
403
XM_005258680.4
XP_005258737.3
4
393
XM_005258681.4
XP_005258738.3
4
393
XM_005258682.4
XP_005258739.3
4
394
XM_005258683.4
XP_005258740.3
4
393
XM_006723116.3
XP_006723179.2
4
394
XM_011526675.2
XP_011524977.1
5
439
XM_011526676.2
XP_011524978.1
5
404
XM_011526677.2
XP_011524979.1
4
393
XM_011526678.2
XP_011524980.1
5
439
XM_011526680.2
XP_011524982.1
4
393
XM_011526682.2
XP_011524984.1
4
410
XM_017026532.1
XP_016882021.1
4
394
XM_017026533.1
XP_016882022.1
4
329
XM_017026534.1
XP_016882023.1
4
302
XM_017026535.1
XP_016882024.1
5
426
GenBank mRNA
AI792400.1
-
5
384
AK301136.1
-
4
364
BC006331.2
-
5
393
BC011914.2
-
5
393
BC028001.1
-
5
404
5198812
BC126293.1
-
5
360
8992009
BC136464.1
-
5
360
9020453
BC144129.1
-
5
360
9052649
DB490139.2
-
5
434
U03858.1
-
4
376
U04806.1
-
5
385
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
signal transduction
positive regulation of cell proliferation
lymphocyte differentiation
regulation of myeloid dendritic cell activation
positive regulation of natural killer cell differentiation
embryonic hemopoiesis
positive regulation of myoblast differentiation
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
homeostasis of number of cells within a tissue
positive regulation of cell proliferation in bone marrow
negative regulation of apoptotic process in bone marrow
positive regulation of osteoclast proliferation
positive regulation of myoblast fusion
receptor binding
cytokine activity
receptor tyrosine kinase binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0