Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C1ORF101 SIRNA Assays ›
Gene Symbol:
C1orf101
Gene Name:
chromosome 1 open reading frame 101
Gene Aliases:
-
Chromosome Location:
Chr.1: 244452014 - 244640362 on Build GRCh38
UniGene ID:
Hs.459534
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS177796
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C1orf101
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130957.1
NP_001124429.1
11
1977
NM_001242340.1
NP_001229269.1
8
1825
NM_173807.4
NP_776168.1
11
1977
XM_011544138.2
XP_011542440.1
11
2028
XM_011544139.2
XP_011542441.1
10
1923
XM_011544142.2
XP_011542444.1
12
2251
XM_011544144.2
XP_011542446.1
10
1954
XM_011544145.2
XP_011542447.1
9
1848
XM_011544149.2
XP_011542451.1
9
1810
XM_011544151.2
XP_011542453.1
9
1812
XM_017000943.1
XP_016856432.1
11
2029
XM_017000944.1
XP_016856433.1
7
2197
XM_017000945.1
XP_016856434.1
8
1820
XM_017000946.1
XP_016856435.1
10
2128
XM_017000947.1
XP_016856436.1
11
2029
XM_017000948.1
XP_016856437.1
12
1973
XM_017000949.1
XP_016856438.1
10
2178
XM_017000950.1
XP_016856439.1
9
1824
XM_017000951.1
XP_016856440.1
11
2029
XM_017000952.1
XP_016856441.1
11
2029
XM_017000953.1
XP_016856442.1
11
2030
GenBank mRNA
AK027205.1
-
8
1464
AK302609.1
-
11
1930
AK303059.1
-
8
1785
BC032859.1
-
11
1962
5269307
BX648685.1
-
7
1678
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0