Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM184B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195071.1 | NP_001182000.1 | 3 | 469 | ||
NM_001195072.1 | NP_001182001.1 | 4 | 555 | |||
NM_012264.4 | NP_036396.2 | 3 | 454 | |||
XM_011530112.2 | XP_011528414.2 | 3 | 503 | |||
XM_011530113.1 | XP_011528415.1 | 3 | 433 | |||
XM_011530114.1 | XP_011528416.1 | 3 | 465 | |||
XM_011530116.2 | XP_011528418.2 | 3 | 495 | |||
XM_011530117.1 | XP_011528419.1 | 4 | 531 | |||
XM_011530118.1 | XP_011528420.1 | 4 | 505 | |||
XM_017028754.1 | XP_016884243.1 | 3 | 504 | |||
XM_017028755.1 | XP_016884244.1 | 3 | 497 | |||
XM_017028756.1 | XP_016884245.1 | 3 | 433 | |||
XM_017028758.1 | XP_016884247.1 | 3 | 433 | |||
GenBank mRNA | AB097053.1 | - | 3 | 425 | ||
AK292652.1 | - | 3 | 454 | |||
AK309071.1 | - | 3 | 425 | |||
AL096879.1 | - | 2 | 147 | |||
BC015489.1 | - | 3 | 410 | 3896682 | ||
BI197128.1 | - | 2 | 148 | |||
BX648345.1 | - | 3 | 505 | |||
CR456413.1 | - | 2 | 288 | |||
CR749859.1 | - | 4 | 461 |