Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSMF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130969.1 | NP_001124441.1 | 6 | 989 | ||
NM_001178064.1 | NP_001171535.1 | 6 | 989 | |||
NM_015537.4 | NP_056352.3 | 5 | 983 | |||
XM_005266061.4 | XP_005266118.1 | 5 | 1006 | |||
GenBank mRNA | AK027474.1 | - | 3 | 254 | ||
AK074602.1 | - | 5 | 951 | |||
AL117660.2 | - | 2 | 101 | |||
AL389944.1 | - | 1 | 37 | |||
AY255128.1 | - | 6 | 989 | |||
AY255129.1 | - | 5 | 983 | |||
AY255131.1 | - | 6 | 989 | |||
AY255132.1 | - | 6 | 989 | |||
BC016862.2 | - | 1 | 30 | |||
BC072412.1 | - | 5 | 968 | 6165009 |