Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FOXP1 SIRNA Assays ›
Gene Symbol:
FOXP1
Gene Name:
forkhead box P1
Gene Aliases:
12CC4, HSPC215, MFH, QRF1, hFKH1B
Chromosome Location:
Chr.3: 70954714 - 71583989 on Build GRCh38
UniGene ID:
Hs.59368
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS178309
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FOXP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001244808.1
NP_001231737.1
20
2366
NM_001244810.1
NP_001231739.1
20
2314
NM_001244812.1
NP_001231741.1
15
1666
NM_001244813.1
NP_001231742.1
14
1558
NM_001244814.1
NP_001231743.1
16
2063
NM_001244815.1
NP_001231744.1
15
2004
NM_001244816.1
NP_001231745.1
19
2171
NM_032682.5
NP_116071.2
20
2266
XM_005264735.3
XP_005264792.1
15
1952
XM_005264736.3
XP_005264793.1
15
2001
XM_005264737.4
XP_005264794.1
20
2272
XM_005264742.3
XP_005264799.1
14
1558
XM_006713102.2
XP_006713165.1
19
2227
XM_006713103.2
XP_006713166.1
17
1964
XM_006713104.2
XP_006713167.1
18
2197
XM_011533584.2
XP_011531886.1
20
2337
XM_011533585.2
XP_011531887.1
19
2189
XM_011533588.2
XP_011531890.1
15
1787
XM_017006165.1
XP_016861654.1
19
2108
XM_017006166.1
XP_016861655.1
19
2204
XM_017006167.1
XP_016861656.1
15
1998
GenBank mRNA
AB052767.1
-
10
1003
AF146696.1
-
18
2003
AF151049.1
-
3
158
AF250920.1
-
19
2171
AF275309.1
-
11
1111
AK027264.1
-
15
1636
AK074321.1
-
16
2062
AK074569.1
-
14
1559
AK092383.1
-
15
1666
AK122710.1
-
20
2366
AK297649.1
-
15
1469
CR457164.1
-
2
66
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of B cell apoptotic process
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
positive regulation of endothelial cell migration
osteoclast differentiation
response to lipopolysaccharide
regulation of tumor necrosis factor production
chemokine (C-C motif) ligand 2 secretion
osteoclast development
macrophage activation
monocyte activation
endothelial cell activation
regulation of monocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of smooth muscle cell proliferation
regulation of interleukin-1 beta secretion
regulation of inflammatory response
negative regulation of androgen receptor signaling pathway
T follicular helper cell differentiation
interleukin-21 secretion
regulation of defense response to bacterium
regulation of macrophage colony-stimulating factor production
regulation of endothelial tube morphogenesis
regulation of interleukin-12 secretion
RNA polymerase II transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding
protein self-association
metal ion binding
androgen receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0