Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FOXP1 SIRNA Assays ›
          Gene Symbol:
          FOXP1
          Gene Name:
          forkhead box P1
          Gene Aliases:
          12CC4, HSPC215, MFH, QRF1, hFKH1B
          Chromosome Location:
          Chr.3: 70954714 - 71583989 on Build GRCh38
          UniGene ID:
          Hs.59368
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS178310
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FOXP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001244808.1 NP_001231737.1 8 935
          NM_001244810.1 NP_001231739.1 8 832
          NM_001244813.1 NP_001231742.1 2 124
          NM_001244814.1 NP_001231743.1 4 629
          NM_001244815.1 NP_001231744.1 3 570
          NM_001244816.1 NP_001231745.1 7 737
          NM_032682.5 NP_116071.2 8 832
          XM_005264735.3 XP_005264792.1 3 521
          XM_005264736.3 XP_005264793.1 3 570
          XM_005264737.4 XP_005264794.1 8 841
          XM_005264742.3 XP_005264799.1 2 127
          XM_006713102.2 XP_006713165.1 7 793
          XM_006713103.2 XP_006713166.1 5 530
          XM_006713104.2 XP_006713167.1 6 763
          XM_011533584.2 XP_011531886.1 8 903
          XM_011533585.2 XP_011531887.1 7 755
          XM_011533588.2 XP_011531890.1 3 353
          XM_017006165.1 XP_016861654.1 7 674
          XM_017006166.1 XP_016861655.1 7 770
          XM_017006167.1 XP_016861656.1 3 570
          GenBank mRNA AF146696.1 - 6 569
          AF250920.1 - 7 737
          AK027264.1 - 3 202
          AK074321.1 - 4 628
          AK122710.1 - 8 935
          BC131720.1 - 5 457
          BX647682.1 - 3 218

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of B cell apoptotic process
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          positive regulation of endothelial cell migration
          osteoclast differentiation
          response to lipopolysaccharide
          regulation of tumor necrosis factor production
          chemokine (C-C motif) ligand 2 secretion
          osteoclast development
          macrophage activation
          monocyte activation
          endothelial cell activation
          regulation of monocyte differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of smooth muscle cell proliferation
          regulation of interleukin-1 beta secretion
          regulation of inflammatory response
          negative regulation of androgen receptor signaling pathway
          T follicular helper cell differentiation
          interleukin-21 secretion
          regulation of defense response to bacterium
          regulation of macrophage colony-stimulating factor production
          regulation of endothelial tube morphogenesis
          regulation of interleukin-12 secretion
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          protein binding
          sequence-specific DNA binding
          protein self-association
          metal ion binding
          androgen receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0