Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APBB2 SIRNA Assays ›
          Gene Symbol:
          APBB2
          Gene Name:
          amyloid beta precursor protein binding family B member 2
          Gene Aliases:
          FE65L, FE65L1
          Chromosome Location:
          Chr.4: 40810027 - 41216714 on Build GRCh38
          UniGene ID:
          Hs.479602
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS179350
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: APBB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166050.1 NP_001159522.1 6 1103
          NM_004307.1 NP_004298.1 6 1103
          NM_173075.4 NP_775098.2 6 1103
          XM_005248101.3 XP_005248158.1 6 1100
          XM_006714005.3 XP_006714068.1 6 1100
          XM_006714007.3 XP_006714070.1 6 1098
          XM_006714008.3 XP_006714071.1 5 1002
          XM_006714009.3 XP_006714072.1 5 1000
          XM_006714010.3 XP_006714073.1 6 1100
          XM_006714011.3 XP_006714074.1 6 1100
          XM_006714012.3 XP_006714075.1 6 1100
          XM_011513686.2 XP_011511988.1 7 1122
          XM_011513687.2 XP_011511989.1 6 1100
          XM_011513688.2 XP_011511990.1 6 1100
          XM_017008142.1 XP_016863631.1 6 1021
          XM_017008143.1 XP_016863632.1 7 1122
          XM_017008144.1 XP_016863633.1 5 1002
          XM_017008145.1 XP_016863634.1 5 1002
          XM_017008146.1 XP_016863635.1 5 1034
          XM_017008147.1 XP_016863636.1 5 1029
          XM_017008148.1 XP_016863637.1 4 933
          XM_017008149.1 XP_016863638.1 5 1034
          XM_017008150.1 XP_016863639.1 6 1021
          XM_017008151.1 XP_016863640.1 5 1002
          XM_017008152.1 XP_016863641.1 5 1002
          XM_017008153.1 XP_016863642.1 6 1021
          XM_017008154.1 XP_016863643.1 5 1002
          XM_017008155.1 XP_016863644.1 5 1002
          XM_017008156.1 XP_016863645.1 5 1034
          XM_017008157.1 XP_016863646.1 4 933
          XM_017008158.1 XP_016863647.1 5 1034
          XM_017008159.1 XP_016863648.1 6 1100
          GenBank mRNA AK226179.1 - 6 1017
          AK295974.1 - 1 441
          AK304247.1 - 6 974
          BC027946.1 - 6 943 5203925
          DA603009.1 - 1 79

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          regulation of transcription, DNA-templated
          cell cycle arrest
          axon guidance
          extracellular matrix organization
          negative regulation of cell growth
          intracellular signal transduction
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          beta-amyloid binding
          protein binding
          transcription factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0