Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDYE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128076.1 | NP_001121548.1 | 2 | 343 | ||
XM_005256632.4 | XP_005256689.1 | 2 | 1212 | |||
XM_005256633.4 | XP_005256690.1 | 2 | 1212 | |||
GenBank mRNA | BC146949.1 | - | 2 | 343 | 9056877 | |
BC146966.1 | - | 2 | 343 | 9056894 |