Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IER5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203434.2 | NP_982258.2 | 1 | 1275 | ||
GenBank mRNA | BC064028.1 | - | 2 | 906 | 6526871 | |
BQ574938.1 | - | 1 | 340 |