Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MID1 SIRNA Assays ›
Gene Symbol:
MID1
Gene Name:
midline 1
Gene Aliases:
BBBG1, FXY, GBBB1, MIDIN, OGS1, OS, OSX, RNF59, TRIM18, XPRF, ZNFXY
Chromosome Location:
Chr.X: 10445310 - 11111264 on Build GRCh38
UniGene ID:
Hs.27695
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS181107
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MID1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000381.3
NP_000372.1
4
1240
NM_001098624.2
NP_001092094.1
4
1170
NM_001193277.1
NP_001180206.1
4
1029
NM_001193278.1
NP_001180207.1
4
1048
NM_001193279.1
NP_001180208.1
3
895
NM_001193280.1
NP_001180209.1
3
781
NM_033289.1
NP_150631.1
4
1126
NM_033290.3
NP_150632.1
4
1032
XM_005274536.1
XP_005274593.1
5
1205
XM_005274537.1
XP_005274594.1
5
1182
XM_006724492.3
XP_006724555.1
5
1804
XM_011545525.2
XP_011543827.1
7
1506
XM_011545526.2
XP_011543828.1
7
1541
XM_011545527.2
XP_011543829.1
5
1451
XM_017029534.1
XP_016885023.1
5
1371
XM_017029535.1
XP_016885024.1
7
1392
XM_017029536.1
XP_016885025.1
6
1468
XM_017029537.1
XP_016885026.1
6
1239
XM_017029538.1
XP_016885027.1
4
938
XM_017029539.1
XP_016885028.1
4
1100
XM_017029540.1
XP_016885029.1
6
1353
XM_017029541.1
XP_016885030.1
7
1506
XM_017029542.1
XP_016885031.1
6
1353
GenBank mRNA
AF035360.1
-
4
926
AF041206.1
-
4
1120
AF041208.1
-
4
1240
AF041210.1
-
3
729
AF230976.1
-
4
1025
AF230977.1
-
4
1025
AF269101.1
-
5
1148
AK025187.1
-
3
211
AK226104.1
-
4
944
AK291215.1
-
4
1122
AK297205.1
-
5
1036
AK299061.1
-
4
1065
AK315095.1
-
4
1071
AY539962.1
-
3
222
AY539963.1
-
4
383
AY539964.1
-
4
375
AY539979.1
-
4
326
AY539980.1
-
3
256
AY539981.1
-
2
103
AY539982.1
-
2
111
AY539983.1
-
3
265
AY539984.1
-
3
265
AY539985.1
-
3
465
AY539986.1
-
2
312
AY539994.1
-
4
992
AY539995.1
-
4
607
AY539996.1
-
4
493
AY539997.1
-
2
139
AY539998.1
-
3
332
AY540014.1
-
3
340
AY540015.1
-
3
454
AY540022.1
-
3
839
AY540023.1
-
3
725
BC053626.1
-
4
1097
6163881
Y13667.1
-
4
1025
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
negative regulation of microtubule depolymerization
pattern specification process
positive regulation of stress-activated MAPK cascade
protein localization to microtubule
interferon-gamma-mediated signaling pathway
protein binding
microtubule binding
zinc ion binding
ligase activity
ubiquitin protein ligase binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
phosphoprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix