Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYLK SIRNA Assays ›
Gene Symbol:
MYLK
Gene Name:
myosin light chain kinase
Gene Aliases:
AAT7, KRP, MLCK, MLCK1, MLCK108, MLCK210, MSTP083, MYLK1, smMLCK
Chromosome Location:
Chr.3: 123612296 - 123884302 on Build GRCh38
UniGene ID:
Hs.477375
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS181411
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYLK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321309.1
NP_001308238.1
33
5662
NM_053025.3
NP_444253.3
34
5870
NM_053026.3
NP_444254.3
33
5663
NM_053027.3
NP_444255.3
33
5717
NM_053028.3
NP_444256.3
32
5510
XM_011512860.2
XP_011511162.1
31
5588
XM_011512861.2
XP_011511163.1
30
5387
XM_017006469.1
XP_016861958.1
17
3146
XM_017006470.1
XP_016861959.1
16
2134
XM_017006471.1
XP_016861960.1
16
2187
XM_017006472.1
XP_016861961.1
4
480
XM_017006473.1
XP_016861962.1
4
429
GenBank mRNA
AB037663.1
-
17
2946
AF069601.2
-
31
5500
AF069602.1
-
31
5551
AF069603.1
-
30
5344
AF069604.1
-
11
1526
AF096773.1
-
3
504
AF096774.1
-
3
501
AF096775.1
-
3
504
AK127437.1
-
1
1393
AK300610.1
-
16
2120
AK314412.1
-
3
515
AK314443.1
-
3
558
AY339601.1
-
31
5588
AY424269.1
-
30
5383
AY424270.1
-
31
5590
BC017811.1
-
3
525
BC064420.1
-
3
544
BC100761.2
-
3
386
BC100762.1
-
3
386
BC100763.1
-
3
383
BC107783.1
-
3
528
BC113456.1
-
3
349
BC113458.1
-
3
346
CR749364.1
-
3
5330
U48959.2
-
32
5707
X85337.1
-
18
3200
X90870.1
-
9
1220
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
protein phosphorylation
muscle contraction
smooth muscle contraction
tonic smooth muscle contraction
positive regulation of cell migration
bleb assembly
positive regulation of calcium ion transport
aorta smooth muscle tissue morphogenesis
cellular hypotonic response
positive regulation of wound healing
mitophagy in response to mitochondrial depolarization
xenophagy
actin binding
myosin light chain kinase activity
protein binding
calmodulin binding
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0