Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024940.2 | NP_001020111.1 | 6 | 1217 | ||
NM_001134855.1 | NP_001128327.1 | 6 | 1369 | |||
NM_016102.3 | NP_057186.1 | 6 | 1253 | |||
XM_006711779.2 | XP_006711842.1 | 6 | 1280 | |||
XM_011544209.2 | XP_011542511.1 | 6 | 1267 | |||
XM_011544210.2 | XP_011542512.1 | 6 | 1369 | |||
XM_011544211.2 | XP_011542513.1 | 6 | 1270 | |||
XM_017001418.1 | XP_016856907.1 | 5 | 1821 | |||
XM_017001419.1 | XP_016856908.1 | 6 | 1365 | |||
GenBank mRNA | AF156271.1 | - | 6 | 976 | ||
AK055426.1 | - | 5 | 1821 | |||
AK301105.1 | - | 6 | 1173 | |||
AK308774.1 | - | 6 | 1244 | |||
AK316379.1 | - | 6 | 1007 | |||
BC033788.1 | - | 6 | 1017 | 5165609 |