Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C21ORF58 SIRNA Assays ›
Gene Symbol:
C21orf58
Gene Name:
chromosome 21 open reading frame 58
Gene Aliases:
-
Chromosome Location:
Chr.21: 46300426 - 46323899 on Build GRCh38
UniGene ID:
Hs.236572
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS182430
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C21orf58
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286462.1
NP_001273391.1
2
345
NM_001286463.1
NP_001273392.1
1
156
NM_001286476.1
NP_001273405.1
2
1418
NM_001286477.1
NP_001273406.1
2
877
NM_058180.4
NP_478060.2
2
1418
XM_005261148.4
XP_005261205.1
2
1404
XM_006724018.3
XP_006724081.1
2
1404
XM_011529619.2
XP_011527921.1
2
1404
XM_011529620.2
XP_011527922.1
2
1404
XM_011529621.2
XP_011527923.1
2
1404
XM_011529623.2
XP_011527925.1
2
3406
XM_011529624.2
XP_011527926.1
1
28
XM_011529625.2
XP_011527927.1
2
555
XM_011529626.2
XP_011527928.1
3
3583
XM_011529627.2
XP_011527929.1
3
3583
XM_011529628.1
XP_011527930.1
2
1404
XM_017028388.1
XP_016883877.1
3
3583
XM_017028389.1
XP_016883878.1
3
3583
XM_017028390.1
XP_016883879.1
3
1581
XM_017028391.1
XP_016883880.1
2
1404
XM_017028392.1
XP_016883881.1
3
402
XM_017028393.1
XP_016883882.1
1
28
GenBank mRNA
AK056387.1
-
2
1391
AK096641.1
-
1
1073
AK098098.1
-
1
93
AY039243.1
-
1
142
AY039244.1
-
1
142
BC026042.1
-
2
1393
BC121007.1
-
3
1565
BC121008.1
-
2
1388
40116957
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0