Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RNF111 SIRNA Assays ›
Gene Symbol:
RNF111
Gene Name:
ring finger protein 111
Gene Aliases:
ARK
Chromosome Location:
Chr.15: 58987588 - 59097419 on Build GRCh38
UniGene ID:
Hs.404423
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS182645
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RNF111
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270528.1
NP_001257457.1
14
3390
NM_001270529.1
NP_001257458.1
14
3360
NM_001270530.1
NP_001257459.1
14
3244
NM_017610.7
NP_060080.6
14
3363
XM_005254479.3
XP_005254536.1
14
3454
XM_006720575.3
XP_006720638.1
15
3625
XM_006720579.3
XP_006720642.1
14
3271
XM_011521694.2
XP_011519996.2
14
3407
XM_011521696.2
XP_011519998.2
14
3077
XM_011521697.2
XP_011519999.2
14
3074
XM_011521699.2
XP_011520001.2
14
3045
XM_017022336.1
XP_016877825.1
15
3493
XM_017022337.1
XP_016877826.1
14
3451
XM_017022338.1
XP_016877827.1
14
3465
XM_017022339.1
XP_016877828.1
14
3427
XM_017022340.1
XP_016877829.1
14
3424
XM_017022341.1
XP_016877830.1
14
3241
XM_017022342.1
XP_016877831.1
14
3217
XM_017022343.1
XP_016877832.1
14
3217
GenBank mRNA
AK095327.1
-
9
1466
AK131304.1
-
14
3245
AK131488.1
-
13
2274
AL157474.1
-
5
590
AL832089.1
-
11
1972
BC010369.1
-
6
590
BC020984.1
-
9
1308
BC060862.1
-
14
3363
30336202
BX538130.1
-
14
3364
BX647259.1
-
14
3104
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
pattern specification process
protein ubiquitination
positive regulation of transforming growth factor beta receptor signaling pathway
ubiquitin-dependent SMAD protein catabolic process
positive regulation of protein ubiquitination
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
global genome nucleotide-excision repair
protein binding
zinc ion binding
ligase activity
SUMO polymer binding
SMAD binding
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0