Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100621.2 | NP_001094091.1 | 3 | 252 | ||
NM_001100622.2 | NP_001094092.1 | 3 | 252 | |||
NM_001288770.1 | NP_001275699.1 | 3 | 413 | |||
NM_001288771.1 | NP_001275700.1 | 3 | 413 | |||
NM_017941.5 | NP_060411.2 | 3 | 252 | |||
XM_005257487.3 | XP_005257544.1 | 3 | 245 | |||
XM_006721966.3 | XP_006722029.1 | 3 | 252 | |||
XM_011524961.1 | XP_011523263.1 | 3 | 252 | |||
XM_011524962.2 | XP_011523264.1 | 2 | 150 | |||
XM_017024806.1 | XP_016880295.1 | 2 | 178 | |||
GenBank mRNA | AK000728.1 | - | 3 | 212 | ||
AK292838.1 | - | 3 | 181 | |||
AK301905.1 | - | 3 | 413 | |||
AL136740.1 | - | 3 | 219 | |||
AY163812.1 | - | 3 | 237 | |||
BC005005.2 | - | 2 | 157 | 3544728 | ||
DC372573.1 | - | 3 | 260 |