Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319956.1 | NP_001306885.1 | 2, 3 | 1099 | ||
NM_018150.3 | NP_060620.2 | 2, 3 | 1103 | |||
XM_005270992.1 | XP_005271049.1 | 2, 3 | 1094 | |||
XM_005270993.1 | XP_005271050.1 | 2, 3 | 1012 | |||
XM_006710735.3 | XP_006710798.1 | 2, 3 | 1271 | |||
XM_011541698.1 | XP_011540000.1 | 2, 3 | 788 | |||
XM_011541699.1 | XP_011540001.1 | 2, 3 | 1052 | |||
XM_011541700.1 | XP_011540002.1 | 2, 3 | 831 | |||
XM_017001623.1 | XP_016857112.1 | 2, 3 | 1052 | |||
XM_017001624.1 | XP_016857113.1 | 2, 3 | 1066 | |||
XM_017001625.1 | XP_016857114.1 | 2, 3 | 810 | |||
GenBank mRNA | AK025744.1 | - | 1, 2 | 192 | ||
AK056424.1 | - | 2, 3 | 1010 | |||
BC034221.1 | - | 2, 3 | 975 | 4814781 | ||
BC098266.1 | - | 2, 3 | 773 | 40026694 | ||
BC098300.1 | - | 2, 3 | 773 | 40026695 |