Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIAA1217 SIRNA Assays ›
Gene Symbol:
KIAA1217
Gene Name:
KIAA1217
Gene Aliases:
SKT
Chromosome Location:
Chr.10: 23694746 - 24547848 on Build GRCh38
UniGene ID:
Hs.445885
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS183463
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIAA1217
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098500.2
NP_001091970.1
19
4254
NM_001282767.1
NP_001269696.1
19
4178
NM_001282768.1
NP_001269697.1
18
4103
NM_001321681.1
NP_001308610.1
14
3168
NM_019590.4
NP_062536.2
21
5885
XM_005252516.3
XP_005252573.1
15
5514
XM_011519552.2
XP_011517854.2
22
5940
XM_011519555.2
XP_011517857.2
21
5858
XM_011519558.2
XP_011517860.2
21
5822
XM_011519559.2
XP_011517861.2
20
5527
XM_011519562.2
XP_011517864.2
20
4263
XM_011519564.2
XP_011517866.1
18
3830
XM_011519565.2
XP_011517867.2
18
3834
XM_011519566.2
XP_011517868.2
18
4044
XM_017016416.1
XP_016871905.1
22
5966
XM_017016417.1
XP_016871906.1
20
5780
XM_017016418.1
XP_016871907.1
20
6602
XM_017016419.1
XP_016871908.1
19
6497
XM_017016420.1
XP_016871909.1
20
5349
XM_017016421.1
XP_016871910.1
22
5559
XM_017016422.1
XP_016871911.1
19
5244
XM_017016423.1
XP_016871912.1
17
4896
XM_017016424.1
XP_016871913.1
17
4968
XM_017016425.1
XP_016871914.1
20
4262
XM_017016426.1
XP_016871915.1
20
4224
XM_017016427.1
XP_016871916.1
19
3930
XM_017016428.1
XP_016871917.1
17
3409
XM_017016429.1
XP_016871918.1
20
4303
GenBank mRNA
AB033043.2
-
19
4254
AB291616.1
-
17
3630
AK055179.1
-
3
1221
AK300422.1
-
17
4520
AL157473.1
-
6
914
AL833280.1
-
21
5513
BC136521.1
-
19
3796
9020511
BC144227.1
-
18
3721
9052747
BX640796.1
-
15
5515
BX648451.1
-
13
3838
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
embryonic skeletal system development
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0