Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTUD7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020205.3 | NP_064590.2 | 12 | 2800 | ||
XM_011509782.1 | XP_011508084.1 | 12 | 2824 | |||
XM_011509783.2 | XP_011508085.1 | 12 | 3054 | |||
XM_011509784.2 | XP_011508086.1 | 12 | 2618 | |||
XM_011509785.2 | XP_011508087.1 | 12 | 6251 | |||
XM_011509786.1 | XP_011508088.1 | 12 | 2690 | |||
XM_011509787.1 | XP_011508089.1 | 12 | 2433 | |||
XM_011509788.1 | XP_011508090.1 | 11 | 2542 | |||
XM_017001850.1 | XP_016857339.1 | 12 | 2663 | |||
XM_017001851.1 | XP_016857340.1 | 11 | 2649 | |||
GenBank mRNA | AJ293573.1 | - | 12 | 2643 | ||
AL122102.1 | - | 2 | 1137 | |||
BC072681.1 | - | 12 | 2737 | 6170513 |