Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MMS19 SIRNA Assays ›
Gene Symbol:
MMS19
Gene Name:
MMS19 homolog, cytosolic iron-sulfur assembly component
Gene Aliases:
MET18, MMS19L, hMMS19
Chromosome Location:
Chr.10: 97458324 - 97498794 on Build GRCh38
UniGene ID:
Hs.500721
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS184710
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MMS19
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289403.1
NP_001276332.1
27
2940
NM_001289404.1
NP_001276333.1
27
3005
NM_001289405.1
NP_001276334.1
29
2951
NM_022362.4
NP_071757.4
28
3069
XM_005270035.2
XP_005270092.1
27
2826
XM_005270041.1
XP_005270098.1
26
2714
XM_006717944.3
XP_006718007.2
28
2860
XM_006717945.2
XP_006718008.1
26
3402
XM_011540062.1
XP_011538364.1
27
2672
XM_011540063.2
XP_011538365.1
26
3400
XM_017016515.1
XP_016872004.1
29
2975
XM_017016516.1
XP_016872005.1
29
2972
XM_017016517.1
XP_016872006.1
28
2846
XM_017016518.1
XP_016872007.1
27
2731
XM_017016519.1
XP_016872008.1
27
2681
XM_017016520.1
XP_016872009.1
26
2569
XM_017016521.1
XP_016872010.1
26
2566
XM_017016522.1
XP_016872011.1
28
2967
XM_017016523.1
XP_016872012.1
27
2851
XM_017016524.1
XP_016872013.1
27
2823
XM_017016525.1
XP_016872014.1
26
2593
XM_017016526.1
XP_016872015.1
27
2829
XM_017016527.1
XP_016872016.1
27
3270
XM_017016528.1
XP_016872017.1
25
3241
XM_017016529.1
XP_016872018.1
27
3470
XM_017016530.1
XP_016872019.1
28
2989
XM_017016531.1
XP_016872020.1
26
2687
XM_017016532.1
XP_016872021.1
25
2464
XM_017016533.1
XP_016872022.1
25
2541
XM_017016534.1
XP_016872023.1
26
3397
XM_017016535.1
XP_016872024.1
25
3258
GenBank mRNA
AF007151.1
-
14
1531
AF319947.1
-
29
2917
AF357881.1
-
28
2868
AJ306408.1
-
28
2851
AK025496.1
-
15
1547
AK027710.1
-
25
2409
AK056244.1
-
17
1876
AK056581.1
-
22
2206
AK225312.1
-
28
2854
AK298995.1
-
28
2907
AK304287.1
-
27
2734
AK314374.1
-
28
2867
BC002692.1
-
17
1831
BC006575.2
-
20
2163
BC009396.2
-
11
1135
BC080532.1
-
27
2784
4889876
BC117129.1
-
28
2867
40125680
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
phosphorelay signal transduction system
DNA metabolic process
DNA repair
nucleotide-excision repair
transcription, DNA-templated
cellular response to DNA damage stimulus
chromosome segregation
response to hormone
iron-sulfur cluster assembly
positive regulation of transcription, DNA-templated
transcription coactivator activity
protein binding
enzyme binding
receptor signaling complex scaffold activity
estrogen receptor binding
protein binding, bridging
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-Offline