Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022842.4 | NP_073753.3 | 2 | 249 | ||
NM_178181.2 | NP_835488.1 | 2 | 249 | |||
XM_011534024.1 | XP_011532326.1 | 2 | 207 | |||
XM_017007070.1 | XP_016862559.1 | 2 | 235 | |||
GenBank mRNA | AF468010.1 | - | 2 | 224 | ||
AK026622.1 | - | 2 | 224 | |||
AK314441.1 | - | 2 | 213 | |||
AY026461.1 | - | 2 | 196 | |||
AY167484.1 | - | 2 | 206 | |||
AY358779.1 | - | 2 | 207 | |||
AY375452.1 | - | 2 | 195 | |||
BC021099.1 | - | 2 | 249 | |||
BC069254.1 | - | 2 | 202 | |||
KU215672.1 | - | 1 | 27 | |||
KU257676.1 | - | 1 | 24 |