Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00202-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026794.1 | 3 | 5364 | |||
GenBank mRNA | CR749234.1 | - | 3 | 5304 |
Gene Symbol: LINC00202-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026795.1 | 3 | 3047 | |||
GenBank mRNA | AK097405.1 | - | 2 | 1214 | ||
AK128027.1 | - | 2 | 5414 |