Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032293.2 | NP_001027464.1 | 11 | 1504 | ||
NM_001098507.1 | NP_001091977.1 | 12 | 1597 | |||
NM_003457.3 | NP_003448.1 | 11 | 1549 | |||
XM_011525185.2 | XP_011523487.1 | 12 | 1729 | |||
XM_017025019.1 | XP_016880508.1 | 12 | 1618 | |||
XM_017025020.1 | XP_016880509.1 | 10 | 1477 | |||
GenBank mRNA | AB209215.1 | - | 12 | 1555 | ||
AF046001.1 | - | 11 | 1603 | |||
AK226172.1 | - | 12 | 1570 | |||
AK291801.1 | - | 11 | 1512 | |||
AK298950.1 | - | 12 | 1401 | |||
AK309979.1 | - | 13 | 1768 | |||
AL834501.1 | - | 9 | 989 | |||
BC002372.2 | - | 11 | 1504 | 2822814 | ||
BC008023.1 | - | 12 | 1263 | 3538197 | ||
BT007365.1 | - | 12 | 1152 |