Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC2HC1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042430.2 | NP_001035895.1 | 2 | 475 | ||
NM_024643.3 | NP_078919.2 | 2 | 475 | |||
XM_005268062.3 | XP_005268119.1 | 2 | 551 | |||
XM_005268063.4 | XP_005268120.1 | 2 | 411 | |||
XM_006720255.3 | XP_006720318.1 | 2 | 411 | |||
XM_011537155.2 | XP_011535457.1 | 2 | 513 | |||
XM_011537156.2 | XP_011535458.1 | 2 | 360 | |||
XM_011537157.2 | XP_011535459.1 | 2 | 411 | |||
XM_017021643.1 | XP_016877132.1 | 2 | 411 | |||
XM_017021644.1 | XP_016877133.1 | 2 | 501 | |||
XM_017021645.1 | XP_016877134.1 | 2 | 345 | |||
GenBank mRNA | AK026746.1 | - | 2 | 410 | ||
AK223359.1 | - | 2 | 411 | |||
AK303882.1 | - | 2 | 413 | |||
BC004259.1 | - | 2 | 366 | 3606108 | ||
BQ061857.1 | - | 2 | 456 |