Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159286.1 | NP_001152758.1 | 10 | 1255 | ||
NM_032019.5 | NP_114408.3 | 11 | 1315 | |||
GenBank mRNA | AF393962.1 | - | 11 | 942 | ||
AF407272.1 | - | 11 | 942 | |||
AF407273.1 | - | 10 | 882 | |||
AF426160.1 | - | 11 | 1295 | |||
AL512711.1 | - | 1 | 19 | |||
AY450395.1 | - | 9 | 792 | |||
BC032049.1 | - | 11 | 1128 | |||
BC125082.1 | - | 6 | 549 | |||
BC125083.1 | - | 11 | 989 | 40117772 | ||
CR456465.1 | - | 11 | 998 |