Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NAV2 SIRNA Assays ›
          Gene Symbol:
          NAV2
          Gene Name:
          neuron navigator 2
          Gene Aliases:
          HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2
          Chromosome Location:
          Chr.11: 19345200 - 20121601 on Build GRCh38
          UniGene ID:
          Hs.502116
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS189695
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NAV2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001111018.1 NP_001104488.1 32 6368
          NM_001111019.2 NP_001104489.1 22 3780
          NM_001244963.1 NP_001231892.1 35 6869
          NM_145117.4 NP_660093.2 32 6692
          NM_182964.5 NP_892009.3 33 6701
          XM_005253214.4 XP_005253271.1 21 8176
          XM_006718364.3 XP_006718427.1 34 7122
          XM_006718365.3 XP_006718428.1 34 7092
          XM_006718366.3 XP_006718429.1 35 6355
          XM_006718367.3 XP_006718430.1 24 8389
          XM_006718368.3 XP_006718431.1 21 8212
          XM_006718369.2 XP_006718432.1 24 3884
          XM_011520444.2 XP_011518746.1 34 7182
          XM_011520445.2 XP_011518747.1 34 7125
          XM_011520446.2 XP_011518748.1 33 7083
          XM_011520447.2 XP_011518749.1 33 7080
          XM_011520448.2 XP_011518750.1 35 7062
          XM_011520449.2 XP_011518751.1 32 7017
          XM_011520450.2 XP_011518752.1 32 6981
          XM_011520451.2 XP_011518753.1 34 6569
          XM_011520452.2 XP_011518754.1 33 6831
          XM_011520453.2 XP_011518755.1 24 3819
          XM_011520454.2 XP_011518756.1 21 3609
          XM_017018520.1 XP_016874009.1 36 6983
          XM_017018521.1 XP_016874010.1 31 6972
          XM_017018522.1 XP_016874011.1 35 6939
          XM_017018523.1 XP_016874012.1 35 7868
          XM_017018524.1 XP_016874013.1 35 6691
          XM_017018525.1 XP_016874014.1 29 5534
          XM_017018526.1 XP_016874015.1 25 6170
          GenBank mRNA AB037840.3 - 33 6702
          AB063115.1 - 32 6368
          AB063116.1 - 32 6661
          AF466143.1 - 32 6407
          AF466144.1 - 33 6416
          AJ488102.1 - 35 6607
          AK001892.1 - 7 904
          AK074287.1 - 22 3779
          AK290458.1 - 16 1982
          BC016054.1 - 14 1685
          BC150290.1 - 33 6702 8860130

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of systemic arterial blood pressure by baroreceptor feedback
          sensory perception of sound
          sensory perception of smell
          locomotory behavior
          optic nerve development
          glossopharyngeal nerve development
          vagus nerve development
          helicase activity
          protein binding
          ATP binding
          heparin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0