Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM116 SIRNA Assays ›
Gene Symbol:
TMEM116
Gene Name:
transmembrane protein 116
Gene Aliases:
-
Chromosome Location:
Chr.12: 111894936 - 112013219 on Build GRCh38
UniGene ID:
Hs.506815
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS189721
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM116
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001193453.1
NP_001180382.1
10
1016
NM_001193531.1
NP_001180460.1
11
1121
NM_001294314.1
NP_001281243.1
13
1338
NM_138341.2
NP_612350.1
10
1057
NR_122119.1
9
918
XM_006719678.3
XP_006719741.1
11
1023
XM_011538946.1
XP_011537248.1
11
975
XM_011538947.1
XP_011537249.1
10
870
XM_011538949.2
XP_011537251.1
11
926
XM_011538950.1
XP_011537252.1
10
901
XM_011538951.2
XP_011537253.1
10
894
XM_011538953.1
XP_011537255.1
13
1311
XM_011538955.1
XP_011537257.1
10
918
XM_011538957.1
XP_011537259.1
12
1232
XM_011538958.1
XP_011537260.1
11
1127
XM_017020188.1
XP_016875677.1
12
1195
XM_017020189.1
XP_016875678.1
11
1083
XM_017020190.1
XP_016875679.1
12
1140
XM_017020191.1
XP_016875680.1
10
1004
XM_017020192.1
XP_016875681.1
12
2645
XM_017020193.1
XP_016875682.1
8
734
XM_017020194.1
XP_016875683.1
9
899
GenBank mRNA
AK074648.1
-
13
1250
AL519857.3
-
9
834
BC000282.1
-
10
936
BC022360.1
-
6
545
BC048796.1
-
11
1013
5193616
BC070135.1
-
10
963
3839474
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix