Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172638.1 | NP_001166109.1 | 2 | 953 | ||
NM_152283.4 | NP_689496.4 | 3 | 1026 | |||
XM_017009710.1 | XP_016865199.1 | 3 | 1056 | |||
XM_017009711.1 | XP_016865200.1 | 3 | 1104 | |||
XM_017009712.1 | XP_016865201.1 | 4 | 1129 | |||
XM_017009713.1 | XP_016865202.1 | 4 | 1177 | |||
XM_017009714.1 | XP_016865203.1 | 3 | 1110 | |||
XM_017009715.1 | XP_016865204.1 | 2 | 1063 | |||
XM_017009716.1 | XP_016865205.1 | 2 | 1355 | |||
XM_017009717.1 | XP_016865206.1 | 2 | 1026 | |||
XM_017009718.1 | XP_016865207.1 | 2 | 1355 | |||
XM_017009719.1 | XP_016865208.1 | 2 | 1355 | |||
XM_017009720.1 | XP_016865209.1 | 2 | 994 | |||
XM_017009721.1 | XP_016865210.1 | 3 | 1036 | |||
GenBank mRNA | AK091376.1 | - | 3 | 1720 | ||
AK091550.1 | - | 1 | 371 | |||
AK295714.1 | - | 3 | 1016 | |||
AK303450.1 | - | 2 | 944 | |||
AL832408.1 | - | 1 | 1113 | |||
BC140761.1 | - | 1 | 332 | 9021629 | ||
BC171780.1 | - | 1 | 332 |