Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFAND4 SIRNA Assays ›
Gene Symbol:
ZFAND4
Gene Name:
zinc finger AN1-type containing 4
Gene Aliases:
ANUBL1
Chromosome Location:
Chr.10: 45614567 - 45672962 on Build GRCh38
UniGene ID:
Hs.659717
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS190233
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFAND4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128324.2
NP_001121796.1
7
2036
NM_001282905.1
NP_001269834.1
8
2486
NM_001282906.1
NP_001269835.1
8
2138
NM_174890.3
NP_777550.2
7
2384
XM_005271842.2
XP_005271899.1
6
1909
XM_005271843.2
XP_005271900.1
6
2076
XM_005271844.3
XP_005271901.1
4
1542
XM_011540366.2
XP_011538668.1
5
1980
XM_011540368.1
XP_011538670.1
4
1425
XM_017016931.1
XP_016872420.1
5
2000
XM_017016932.1
XP_016872421.1
7
2146
XM_017016933.1
XP_016872422.1
7
2013
XM_017016934.1
XP_016872423.1
7
2066
XM_017016935.1
XP_016872424.1
6
1816
XM_017016936.1
XP_016872425.1
5
2156
XM_017016937.1
XP_016872426.1
5
2004
XM_017016938.1
XP_016872427.1
5
2298
XM_017016939.1
XP_016872428.1
5
1869
XM_017016940.1
XP_016872429.1
8
2562
XM_017016941.1
XP_016872430.1
8
2105
XM_017016942.1
XP_016872431.1
6
2094
XM_017016943.1
XP_016872432.1
6
2370
XM_017016944.1
XP_016872433.1
6
1927
XM_017016945.1
XP_016872434.1
4
1514
XM_017016946.1
XP_016872435.1
8
2180
GenBank mRNA
AF311324.1
-
7
2000
AK097504.1
-
4
1504
AK292420.1
-
8
2087
AK292469.1
-
7
1985
AK314395.1
-
7
1996
BC045587.1
-
7
2036
5269483
BC048968.1
-
8
2028
5272133
Back To Top
More Information
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix