Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164804.1 | NP_001158276.1 | 9 | 1167 | ||
NM_178893.4 | NP_849224.2 | 10 | 1392 | |||
GenBank mRNA | AK030097.1 | - | 10 | 1395 | ||
AK030971.1 | - | 9 | 1124 | |||
AK054394.1 | - | 9 | 1168 | |||
AK151197.1 | - | 2 | 184 | |||
AK152038.1 | - | 2 | 184 | |||
AK152105.1 | - | 2 | 184 | |||
AK152490.1 | - | 9 | 1156 | |||
AK153134.1 | - | 9 | 1156 | |||
AK154247.1 | - | 9 | 1124 | |||
AK170184.1 | - | 9 | 1125 | |||
BC026634.1 | - | 1 | 102 | |||
BC031164.1 | - | 7 | 814 | |||
BC068206.1 | - | 1 | 544 | |||
BC138731.1 | - | 9 | 1192 | 8861753 | ||
BC138732.1 | - | 9 | 1192 | 8861754 |