Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam210a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153794.4 | NP_722489.1 | 4 | 932 | ||
GenBank mRNA | AK034980.1 | - | 4 | 814 | ||
AK044238.1 | - | 4 | 820 | |||
AK049567.1 | - | 4 | 820 | |||
AK077093.1 | - | 4 | 933 | |||
AK079137.1 | - | 1 | 54 | |||
AK142190.1 | - | 4 | 804 | |||
AK147662.1 | - | 4 | 807 | |||
AK159319.1 | - | 4 | 783 | |||
AK162396.1 | - | 4 | 817 | |||
BC038695.1 | - | 4 | 699 | 1548421 | ||
BC095936.1 | - | 4 | 782 | 3968496 |