Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABCA1 SIRNA Assays ›
          Gene Symbol:
          Abca1
          Gene Name:
          ATP-binding cassette, sub-family A (ABC1), member 1
          Gene Aliases:
          ABC-1, Abc1
          Chromosome Location:
          Chr.4: 53030789 - 53159988 on Build GRCm38
          UniGene ID:
          Mm.277376
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS201491
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Abca1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013454.3 NP_038482.3 32 4818
          GenBank mRNA BP761367.1 - 2 203
          X75926.1 - 31 4581

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          peptide secretion
          protein lipidation
          transport
          phagocytosis, engulfment
          lysosome organization
          G-protein coupled receptor signaling pathway
          cholesterol metabolic process
          positive regulation of cholesterol efflux
          phospholipid transport
          endosomal transport
          cholesterol transport
          positive regulation of cAMP biosynthetic process
          intracellular cholesterol transport
          regulation of Cdc42 protein signal transduction
          cholesterol efflux
          phospholipid efflux
          high-density lipoprotein particle assembly
          apolipoprotein A-I-mediated signaling pathway
          lipoprotein metabolic process
          lipoprotein biosynthetic process
          cholesterol homeostasis
          reverse cholesterol transport
          phospholipid translocation
          interleukin-1 beta secretion
          transmembrane transport
          phospholipid homeostasis
          response to high density lipoprotein particle
          platelet dense granule organization
          cellular response to lipopolysaccharide
          cellular response to retinoic acid
          anion transmembrane transport
          nucleotide binding
          receptor binding
          transporter activity
          protein binding
          ATP binding
          phospholipid transporter activity
          anion transmembrane transporter activity
          ATPase activity
          cholesterol transporter activity
          syntaxin binding
          small GTPase binding
          apolipoprotein binding
          apolipoprotein A-I binding
          apolipoprotein A-I receptor activity
          ATPase activity, coupled to transmembrane movement of substances
          ATPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0