Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198790.1 | NP_001185719.1 | 4 | 229 | ||
NM_001198791.1 | NP_001185720.1 | 4 | 211 | |||
NM_001198792.1 | NP_001185721.1 | 3 | 170 | |||
NM_021515.3 | NP_067490.1 | 3 | 188 | |||
GenBank mRNA | AJ010108.1 | - | 3 | 94 | ||
AJ010109.1 | - | 3 | 184 | |||
AK046613.1 | - | 4 | 230 | |||
AK089270.1 | - | 4 | 212 | |||
BC014802.1 | - | 4 | 155 | 4505142 | ||
BC054366.1 | - | 3 | 177 | 5346232 | ||
BY130582.1 | - | 4 | 212 | |||
BY333336.1 | - | 3 | 170 | |||
CF950379.1 | - | 3 | 188 | |||
DQ486026.1 | - | 4 | 242 |