Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ivns1abp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039511.1 | NP_001034600.1 | 4, 5 | 790 | ||
NM_001039512.1 | NP_001034601.1 | 4, 5 | 790 | |||
NM_054102.2 | NP_473443.2 | 4, 5 | 790 | |||
GenBank mRNA | AB055737.1 | - | 4, 5 | 786 | ||
AB055738.1 | - | 4, 5 | 786 | |||
AK007291.1 | - | 4, 5 | 740 | |||
AK129229.1 | - | 4, 5 | 728 | |||
AK144619.1 | - | 4, 5 | 769 | |||
AK144835.1 | - | 4, 5 | 771 | |||
AK145071.1 | - | 4, 5 | 781 | |||
AK145845.1 | - | 4, 5 | 740 | |||
AK146001.1 | - | 4, 5 | 771 | |||
AK146329.1 | - | 1, 2 | 114 | |||
AK146494.1 | - | 4, 5 | 743 | |||
AK146645.1 | - | 4, 5 | 741 | |||
AK159254.1 | - | 4, 5 | 772 | |||
AK179875.1 | - | 3, 4 | 342 | |||
AK181493.1 | - | 3, 4 | 367 | |||
BC004092.1 | - | 3, 4 | 490 | 3593827 | ||
BC040250.1 | - | 4, 5 | 782 | |||
DQ914522.1 | - | 4, 5 | 742 | |||
DQ914523.1 | - | 4, 5 | 712 |