Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATP2A2 SIRNA Assays ›
          Gene Symbol:
          Atp2a2
          Gene Name:
          ATPase, Ca++ transporting, cardiac muscle, slow twitch 2
          Gene Aliases:
          9530097L16Rik, D5Wsu150e, SERCA2, SERCA2B, Serca2a, mKIAA4195
          Chromosome Location:
          Chr.5: 122453513 - 122502225 on Build GRCm38
          UniGene ID:
          Mm.227583
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS202249
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Atp2a2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001110140.3 NP_001103610.1 13 2153
          NM_009722.3 NP_033852.1 13 2153
          GenBank mRNA AF039893.1 - 9 1300
          AJ131821.1 - 13 1823
          AJ223584.1 - 13 1823
          AK220538.1 - 13 2102
          BC054531.1 - 13 1812 6406916
          BC054748.1 - 13 1812 6406916

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of the force of heart contraction
          transport
          ion transport
          calcium ion transport
          cellular calcium ion homeostasis
          regulation of muscle contraction
          ER-nucleus signaling pathway
          organelle organization
          regulation of cardiac muscle contraction by calcium ion signaling
          transition between fast and slow fiber
          endoplasmic reticulum calcium ion homeostasis
          positive regulation of endoplasmic reticulum calcium ion concentration
          T-tubule organization
          response to peptide hormone
          negative regulation of heart contraction
          relaxation of cardiac muscle
          sarcoplasmic reticulum calcium ion transport
          calcium ion transmembrane transport
          regulation of cardiac muscle cell membrane potential
          regulation of cardiac muscle cell action potential involved in regulation of contraction
          regulation of calcium ion-dependent exocytosis of neurotransmitter
          calcium ion transport from cytosol to endoplasmic reticulum
          calcium ion import into sarcoplasmic reticulum
          nucleotide binding
          calcium-transporting ATPase activity
          calcium ion binding
          protein binding
          ATP binding
          protein C-terminus binding
          hydrolase activity
          enzyme binding
          lutropin-choriogonadotropic hormone receptor binding
          S100 protein binding
          metal ion binding
          calcium-transporting ATPase activity involved in regulation of cardiac muscle cell membrane potential

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66756dbf47-27kf9:80/100.66.130.62:80.
          git-commit: ed967532dff6834307dac93af002e5c8e22ba7c9
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline