Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Runx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145920.2 | NP_001139392.1 | 2 | 1507 | ||
NM_001146038.2 | NP_001139510.1 | 4 | 772 | |||
NM_001271627.1 | NP_001258556.1 | 3 | 936 | |||
NM_001271630.1 | NP_001258559.1 | 2 | 1035 | |||
NM_001271631.1 | NP_001258560.1 | 2 | 1035 | |||
NM_009820.5 | NP_033950.2 | 4 | 739 | |||
GenBank mRNA | AF005936.1 | - | 3 | 890 | ||
AF010284.1 | - | 4 | 739 | |||
AF053954.1 | - | 3 | 353 | |||
AF053955.1 | - | 2 | 187 | |||
AF053956.1 | - | 2 | 225 | |||
AK134925.1 | - | 2 | 293 | |||
AK137061.1 | - | 1 | 118 | |||
AK158925.1 | - | 2 | 1035 | |||
D14636.1 | - | 2 | 1505 | |||
D14637.1 | - | 2 | 1505 | |||
DQ458792.1 | - | 3 | 524 |