Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001026214.2 | NP_001021385.1 | 4 | 446 | ||
NM_001286049.1 | NP_001272978.1 | 3 | 421 | |||
NM_001286058.1 | NP_001272987.1 | 2 | 348 | |||
NM_007647.3 | NP_031673.2 | 3 | 373 | |||
GenBank mRNA | AF006482.1 | - | 3 | 381 | ||
AF136571.1 | - | 3 | 407 | |||
AJ238636.1 | - | 3 | 409 | |||
AK002618.1 | - | 3 | 346 | |||
AK031581.1 | - | 4 | 426 | |||
AK036641.1 | - | 4 | 423 | |||
AK037736.1 | - | 3 | 342 | |||
AK045828.1 | - | 3 | 359 | |||
AK079267.1 | - | 2 | 270 | |||
AK080265.1 | - | 4 | 433 | |||
AK081435.1 | - | 4 | 411 | |||
AK088455.1 | - | 3 | 402 | |||
AK160950.1 | - | 1 | 267 | |||
AK163688.1 | - | 3 | 347 | |||
AW012371.1 | - | 3 | 421 | |||
BC015247.1 | - | 2 | 331 | 4242560 |