Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007711.3 | NP_031737.1 | 8 | 1889 | ||
NM_173873.1 | NP_776298.1 | 9 | 2065 | |||
NM_173874.1 | NP_776299.1 | 9 | 2065 | |||
NM_173876.3 | NP_776301.1 | 8 | 1610 | |||
GenBank mRNA | AF029347.1 | - | 8 | 1513 | ||
AK141136.1 | - | 9 | 2003 | |||
AK147364.1 | - | 8 | 2045 | |||
AK147548.1 | - | 8 | 2047 | |||
AK147621.1 | - | 8 | 2047 | |||
AK149276.1 | - | 9 | 1604 | |||
AK150688.1 | - | 9 | 1994 | |||
AK159698.1 | - | 9 | 2039 | |||
AK161327.1 | - | 9 | 1993 | |||
AK162639.1 | - | 9 | 2006 | |||
AK169297.1 | - | 9 | 1986 | |||
AK169516.1 | - | 3 | 1941 | |||
BC057133.1 | - | 8 | 1543 | 6833732 | ||
BC057855.1 | - | 8 | 1705 | |||
X78874.1 | - | 8 | 1714 |