Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007711.3 | NP_031737.1 | 9, 10 | 2127 | ||
NM_173873.1 | NP_776298.1 | 10, 11 | 2303 | |||
NM_173874.1 | NP_776299.1 | 10, 11 | 2303 | |||
NM_173876.3 | NP_776301.1 | 9, 10 | 1848 | |||
GenBank mRNA | AF029347.1 | - | 9, 10 | 1751 | ||
AK141136.1 | - | 10, 11 | 2241 | |||
AK149276.1 | - | 10, 11 | 1842 | |||
AK150688.1 | - | 10, 11 | 2232 | |||
AK159698.1 | - | 10, 11 | 2277 | |||
AK161327.1 | - | 10, 11 | 2231 | |||
AK162639.1 | - | 10, 11 | 2244 | |||
AK169297.1 | - | 10, 11 | 2224 | |||
BC057133.1 | - | 9, 10 | 1781 | 6833732 | ||
BC057855.1 | - | 9, 10 | 1943 | |||
X78874.1 | - | 9, 10 | 1952 |