Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113389.1 | NP_001106860.1 | 3 | 169 | ||
NM_007890.2 | NP_031916.1 | 3 | 428 | |||
GenBank mRNA | AK140323.1 | - | 3 | 625 | ||
BC129888.1 | - | 2 | 58 | 40111469 | ||
BC129889.1 | - | 2 | 58 | 40111470 | ||
BC138716.1 | - | 2 | 58 | 8861738 | ||
BC145405.1 | - | 2 | 58 | 9053963 | ||
CB526389.1 | - | 3 | 169 | |||
CD540384.1 | - | 3 | 144 | |||
U58497.1 | - | 2 | 58 |