Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB560752.1 | - | 6 | 1429 | ||
AB560757.1 | - | 2 | 486 | |||
AB560758.1 | - | 2 | 212 | |||
AB560759.1 | - | 2 | 210 | |||
AB560760.1 | - | 2 | 162 | |||
AF128220.1 | - | 2 | 419 | |||
AF128221.1 | - | 2 | 419 | |||
AK036627.1 | - | 6 | 1403 | |||
AK041525.1 | - | 7 | 1551 | |||
AK054182.1 | - | 6 | 1434 | |||
AK077236.1 | - | 6 | 1409 | |||
AK136228.1 | - | 2 | 208 | |||
EU791538.1 | - | 6 | 1347 | |||
EU791540.1 | - | 6 | 1472 | |||
M38651.1 | - | 6 | 1411 |