Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015736.2 | NP_056551.2 | 7 | 1613 | ||
GenBank mRNA | AK019995.1 | - | 7 | 1612 | ||
AK135489.1 | - | 7 | 1574 | |||
BC043331.1 | - | 7 | 1469 | 5342768 | ||
U70538.1 | - | 7 | 1457 |