Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008112.4 | NP_032138.3 | 10, 11 | 1411 | ||
GenBank mRNA | AK007871.1 | - | 10, 11 | 1354 | ||
AK078144.1 | - | 1354 | ||||
AK079690.1 | - | 9, 10 | 1252 | |||
AK090083.1 | - | 10, 11 | 1359 | |||
AK137761.1 | - | 10, 11 | 1358 | |||
AK143633.1 | - | 10, 11 | 1356 | |||
AK143681.1 | - | 10, 11 | 1390 | |||
AK149958.1 | - | 10, 11 | 1364 | |||
AK150356.1 | - | 10, 11 | 1346 | |||
AK150655.1 | - | 10, 11 | 1381 | |||
AK152250.1 | - | 10, 11 | 1345 | |||
AK153381.1 | - | 10, 11 | 1324 | |||
AK159734.1 | - | 10, 11 | 1356 | |||
AK166317.1 | - | 10, 11 | 1356 | |||
AK167659.1 | - | 10, 11 | 1368 | |||
AK169299.1 | - | 10, 11 | 1322 | |||
BC024971.1 | - | 10, 11 | 1332 | |||
BC053381.1 | - | 3511 | ||||
BC055341.1 | - | 10, 11 | 1337 | 6808475 | ||
L36314.1 | - | 10, 11 | 1217 |