Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277290.1 | NP_001264219.1 | 8, 9 | 1757 | ||
NM_008238.2 | NP_032264.1 | 8, 9 | 1760 | |||
GenBank mRNA | BC108980.1 | - | 7, 8 | 1645 | 40047616 | |
BC108981.1 | - | 7, 8 | 1648 | 40047617 | ||
X81593.1 | - | 8, 9 | 1710 |