Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013558.2 | NP_038586.2 | 2 | 680 | ||
GenBank mRNA | BC129894.1 | - | 2 | 549 | 40111516 | |
BC129895.1 | - | 2 | 551 | 40111517 | ||
D85732.1 | - | 2 | 651 | |||
EU622851.1 | - | 1 | 507 | |||
M32218.1 | - | 1 | 507 |
Gene Symbol: Gm10501 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK134269.1 | - | 2 | 1603 |